Assay Details
| Gene Symbol: PIF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015291325.2 | XP_015146811.1 | 7 - 8 | 1272 | 65 | |
| XM_426648.6 | XP_426648.4 | 7 - 8 | 1254 | 65 | ||
| GenBank mRNA | CR386691.1 | - | 6 - 7 | 668 | 65 | |
More Information
Set Membership: |
|

PIF1
Genomic Map