Assay Details
| Gene Symbol: TRUB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015288931.2 | XP_015144417.1 | 5 - 6 | 511 | 61 | |
| XM_421776.6 | XP_421776.1 | 7 - 8 | 683 | 61 | ||
| GenBank mRNA | CR387253.1 | - | 5 - 6 | 495 | 61 | |
| CR389445.1 | - | 7 - 8 | 636 | 61 | ||
More Information
Set Membership: |
|

TRUB1
Genomic Map