Assay Details
| Gene Symbol: GLUD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015288033.2 | XP_015143519.1 | 9 - 10 | 1301 | 136 | |
| GenBank mRNA | CR387697.1 | - | 9 - 10 | 1149 | 136 | |
| MF415491.1 | - | 9 - 10 | 1076 | 136 | ||
More Information
Set Membership: |
|

GLUD2
Genomic Map