Assay Details
| Gene Symbol: MYL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001271929.1 | NP_001258858.1 | 4 - 5 | 334 | 92 | |
| GenBank mRNA | BU257651.1 | - | 4 - 5 | 339 | 92 | |
| CR387719.1 | - | 4 - 5 | 478 | 92 | ||
| CR391086.1 | - | 3 - 4 | 294 | 92 | ||
More Information
Set Membership: |
|

MYL2
Genomic Map