Assay Details
| Gene Symbol: OVOB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015286750.2 | XP_015142236.2 | 6 - 7 | 935 | 77 | |
| GenBank mRNA | AB180913.1 | - | 6 - 7 | 842 | 77 | |
More Information
Set Membership: |
|

OVOB
Genomic Map