Assay Details
| Gene Symbol: C3H6ORF203 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001278010.1 | NP_001264939.1 | 2 - 3 | 506 | 86 | |
| GenBank mRNA | BU427211.1 | - | 1 - 2 | 172 | 86 | |
| BU433538.1 | - | 2 - 3 | 506 | 86 | ||
| BU479967.1 | - | 1 - 2 | 217 | 86 | ||
| BX930326.2 | - | 2 - 3 | 480 | 86 | ||
| BX932682.1 | - | 2 - 3 | 502 | 86 | ||
| BX950322.2 | - | 2 - 3 | 501 | 86 | ||
More Information
Set Membership: |
|

C3H6ORF203
Genomic Map