Assay Details
| Gene Symbol: MED8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001277568.1 | NP_001264497.1 | 2 - 3 | 162 | 67 | |
| GenBank mRNA | BX932080.1 | - | 1 - 2 | 135 | 67 | |
| BX934369.2 | - | 1 - 2 | 132 | 67 | ||
| BX936148.1 | - | 1 - 2 | 132 | 67 | ||
| DT659918.1 | - | 2 - 3 | 162 | 67 | ||
More Information
Set Membership: |
|

MED8
Genomic Map