Assay Details
| Gene Symbol: HOMER3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004948900.3 | XP_004948957.1 | 18 - 19 | 6132 | 66 | |
| XM_015300131.2 | XP_015155617.1 | 8 - 9 | 1042 | 66 | ||
| XM_418233.6 | XP_418233.1 | 7 - 8 | 912 | 66 | ||
| GenBank mRNA | CR386688.1 | - | 4 - 5 | 485 | 66 | |
More Information
Set Membership: |
|

HOMER3
Genomic Map