Assay Details
| Gene Symbol: B3GALNT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015291773.2 | XP_015147259.1 | 2 - 3 | 417 | 64 | |
| XM_025153493.1 | XP_025009261.1 | 1 - 2 | 3762 | 64 | ||
| XM_025153494.1 | XP_025009262.1 | 4 - 5 | 643 | 64 | ||
| XM_025153495.1 | XP_025009263.1 | 3 - 4 | 501 | 64 | ||
| XM_025153498.1 | XP_025009266.1 | 2 - 3 | 208 | 64 | ||
| XM_025153501.1 | XP_025009269.1 | 3 - 4 | 676 | 64 | ||
| GenBank mRNA | CR387037.1 | - | 3 - 4 | 505 | 64 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015291773.2, XM_025153493.1, XM_025153494.1, XM_025153495.1, XM_025153498.1 transcripts.
Set Membership: |
|

B3GALNT1
Genomic Map