Assay Details
| Gene Symbol: SPHKAP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001177532.3 | NP_001171003.1 | 7 | 3965 | 80 | |
| XM_015276888.2 | XP_015132374.1 | 7 | 4120 | 80 | ||
| XM_015276889.2 | XP_015132375.1 | 7 | 4118 | 80 | ||
| XM_015276890.1 | XP_015132376.1 | 7 | 3963 | 80 | ||
| XM_015276891.1 | XP_015132377.1 | 7 | 3960 | 80 | ||
| XM_015276892.2 | XP_015132378.1 | 7 | 3958 | 80 | ||
| XM_015276893.2 | XP_015132379.1 | 7 | 3963 | 80 | ||
| XM_015276894.2 | XP_015132380.1 | 7 | 4133 | 80 | ||
| GenBank mRNA | CR387126.1 | - | 1 | 955 | 80 | |
More Information
Set Membership: |
|

SPHKAP
Genomic Map