Assay Details
| Gene Symbol: CCDC189 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004945692.3 | XP_004945749.1 | 3 - 4 | 760 | 60 | |
| XM_004945696.3 | XP_004945753.1 | 4 - 5 | 462 | 60 | ||
| XM_004945697.2 | XP_004945754.1 | 3 - 4 | 644 | 60 | ||
| XM_004945699.3 | XP_004945756.1 | 4 - 5 | 466 | 60 | ||
| XM_025141456.1 | XP_024997224.1 | 3 - 4 | 644 | 60 | ||
| XM_025141458.1 | XP_024997226.1 | 4 - 5 | 432 | 60 | ||
| XM_025141459.1 | XP_024997227.1 | 4 - 5 | 370 | 60 | ||
| XM_025141460.1 | XP_024997228.1 | 3 - 4 | 366 | 60 | ||
| XM_025141461.1 | XP_024997229.1 | 3 - 4 | 366 | 60 | ||
| XM_025141462.1 | XP_024997230.1 | 2 - 3 | 526 | 60 | ||
| GenBank mRNA | CR354343.1 | - | 4 - 5 | 367 | 60 | |
| CR385719.1 | - | 3 - 4 | 250 | 60 | ||
| CR386406.1 | - | 2 - 3 | 711 | 60 | ||
| CR387157.1 | - | 4 - 5 | 431 | 60 | ||
| CR390026.1 | - | 3 - 4 | 253 | 60 | ||
| CR390582.1 | - | 4 - 5 | 269 | 60 | ||
| CR407096.1 | - | 4 - 5 | 422 | 60 | ||
| CR523308.1 | - | 3 - 4 | 644 | 60 | ||
More Information
Set Membership: |
|

CCDC189
Genomic Map