Assay Details
| Gene Symbol: IL15RA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015286682.2 | XP_015142168.1 | 4 - 5 | 570 | 69 | |
| XM_025153116.1 | XP_025008884.1 | 4 - 5 | 572 | 69 | ||
| XM_025153118.1 | XP_025008886.1 | 4 - 5 | 572 | 69 | ||
| GenBank mRNA | CR387487.1 | - | 3 - 4 | 266 | 69 | |
More Information
Set Membership: |
|

IL15RA
Genomic Map