Assay Details
| Gene Symbol: BDH2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015285615.2 | XP_015141101.1 | 7 - 8 | 597 | 73 | |
| XM_420669.6 | XP_420669.1 | 7 - 8 | 601 | 73 | ||
| GenBank mRNA | BX933679.2 | - | 6 - 7 | 549 | 73 | |
| BX933695.2 | - | 6 - 7 | 549 | 73 | ||
| CR387508.1 | - | 6 - 7 | 549 | 73 | ||
More Information
Set Membership: |
|

BDH2
Genomic Map