Assay Details
| Gene Symbol: MSTO1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_422860.6 | XP_422860.2 | 7 - 8 | 626 | 92 | |
| GenBank mRNA | CR386904.1 | - | 4 - 5 | 295 | 92 | |
| CR387885.1 | - | 1 - 2 | 137 | 92 | ||
More Information
Set Membership: |
|

MSTO1
Genomic Map