Assay Details
| Gene Symbol: IRAG2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001348560.1 | NP_001335489.1 | 5 - 6 | 1045 | 69 | |
| GenBank mRNA | CR388830.1 | - | 5 - 6 | 499 | 69 | |
More Information
Set Membership: |
|

IRAG2
Genomic Map