Assay Details
| Gene Symbol: PCDH7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_025149784.1 | XP_025005552.1 | 1 - 2 | 5390 | 57 | |
| GenBank mRNA | CR388971.1 | - | 1 - 2 | 599 | 57 | |
More Information
Set Membership: |
|

PCDH7
Genomic Map