Assay Details
| Gene Symbol: GEN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004935777.3 | XP_004935834.1 | 16 | 3211 | 117 | |
| XM_004935779.3 | XP_004935836.1 | 17 | 3352 | 117 | ||
| XM_004935780.3 | XP_004935837.1 | 14 | 2957 | 117 | ||
| XM_025148850.1 | XP_025004618.1 | 16 | 3271 | 117 | ||
| XM_419963.6 | XP_419963.3 | 15 | 3172 | 117 | ||
| GenBank mRNA | CR389136.1 | - | 1 | 551 | 117 | |
More Information
Set Membership: |
|

GEN1
Genomic Map