Assay Details
| Gene Symbol: ACYP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001168005.2 | NP_001161477.1 | 1 - 2 | 122 | 109 | |
| GenBank mRNA | CR389436.1 | - | 1 - 2 | 92 | 109 | |
More Information
Set Membership: |
|

ACYP1
Genomic Map