Assay Details
Gene Symbol: ZBTB6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277641.2 | NP_001264570.2 | 1 | 1365 | 95 | |
GenBank mRNA | CR389786.1 | - | 1 | 1147 | 95 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
