Assay Details
| Gene Symbol: SMPDL3B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001252280.1 | NP_001239209.1 | 7 | 2220 | 71 | |
| XM_015297553.2 | XP_015153039.1 | 7 | 2210 | 71 | ||
| GenBank mRNA | CR390113.1 | - | 1 | 755 | 71 | |
More Information
Set Membership: |
|

SMPDL3B
Genomic Map