Assay Details
| Gene Symbol: RPS8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001252126.1 | NP_001239055.1 | 3 - 4 | 263 | 98 | |
| GenBank mRNA | AY389966.1 | - | 2 - 3 | 182 | 98 | |
| BX930826.1 | - | 2 - 3 | 215 | 98 | ||
| CN218040.1 | - | 2 - 3 | 195 | 98 | ||
| CR390133.1 | - | 2 - 3 | 222 | 98 | ||
| CR390136.1 | - | 2 - 3 | 217 | 98 | ||
| CR390875.1 | - | 3 - 4 | 252 | 98 | ||
More Information
Set Membership: |
|

RPS8
Genomic Map