Assay Details
| Gene Symbol: RPS8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001252126.1 | NP_001239055.1 | 5 - 6 | 569 | 68 | |
| GenBank mRNA | AY389966.1 | - | 4 - 5 | 488 | 68 | |
| BX930826.1 | - | 4 - 5 | 488 | 68 | ||
| CF250737.1 | - | 5 - 6 | 571 | 68 | ||
| CN218040.1 | - | 4 - 5 | 501 | 68 | ||
| CR390133.1 | - | 4 - 5 | 528 | 68 | ||
| CR390136.1 | - | 4 - 5 | 523 | 68 | ||
| CR390875.1 | - | 5 - 6 | 558 | 68 | ||
More Information
Set Membership: |
|

RPS8
Genomic Map