Assay Details
Gene Symbol: GRM7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004944718.3 | XP_004944775.1 | 2 - 3 | 833 | 62 | |
XM_015293175.2 | XP_015148661.1 | 2 - 3 | 834 | 62 | ||
GenBank mRNA | CR391191.1 | - | 2 - 3 | 241 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
