Assay Details
| Gene Symbol: LOC424577 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_422416.6 | XP_422416.3 | 3 | 2318 | 128 | |
| GenBank mRNA | CR391293.1 | - | 1 | 749 | 129 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC424577| Gene Symbol: LOC424577 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_422416.6 | XP_422416.3 | 3 | 2318 | 128 | |
| GenBank mRNA | CR391293.1 | - | 1 | 749 | 129 | |
Set Membership: |
|