Assay Details
| Gene Symbol: KIF2C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015291291.2 | XP_015146777.1 | 9 - 10 | 900 | 72 | |
| XM_015291292.2 | XP_015146778.1 | 9 - 10 | 829 | 72 | ||
| XM_015291293.2 | XP_015146779.1 | 8 - 9 | 808 | 72 | ||
| XM_015291294.2 | XP_015146780.1 | 8 - 9 | 857 | 72 | ||
| XM_015291295.2 | XP_015146781.1 | 8 - 9 | 789 | 72 | ||
| XM_015291296.2 | XP_015146782.1 | 8 - 9 | 1042 | 72 | ||
| XM_015291297.2 | XP_015146783.1 | 7 - 8 | 733 | 72 | ||
| XM_015291299.2 | XP_015146785.1 | 7 - 8 | 740 | 72 | ||
| XM_015291300.2 | XP_015146786.1 | 7 - 8 | 993 | 72 | ||
| XM_025153279.1 | XP_025009047.1 | 9 - 10 | 823 | 72 | ||
| XM_422422.6 | XP_422422.4 | 9 - 10 | 906 | 72 | ||
| GenBank mRNA | BX930594.2 | - | 7 - 8 | 702 | 72 | |
| BX931004.2 | - | 7 - 8 | 733 | 72 | ||
| BX933284.2 | - | 9 - 10 | 805 | 72 | ||
| CR405925.1 | - | 8 - 9 | 800 | 72 | ||
More Information
Set Membership: |
|

KIF2C
Genomic Map