Assay Details
| Gene Symbol: LOC416146 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | CR406019.1 | - | 1 - 2 | 130 | 113 | |
| Gene Symbol: LOC416147 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_025155025.1 | XP_025010793.1 | 1 - 2 | 842 | 113 | |
| XM_025155026.1 | XP_025010794.1 | 1 - 2 | 842 | 113 | ||
| XM_414478.6 | XP_414478.5 | 1 - 2 | 840 | 113 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_025155025.1, XM_025155026.1, XM_414478.6 transcripts.
Set Membership: |
|

LOC416146
Genomic Map