Assay Details
| Gene Symbol: DNAJC5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001278008.2 | NP_001264937.1 | 3 - 4 | 326 | 100 | |
| NM_001278009.1 | NP_001264938.1 | 3 - 4 | 226 | 100 | ||
| GenBank mRNA | AJ395662.1 | - | 3 - 4 | 226 | 100 | |
| BX934729.2 | - | 1 - 2 | 102 | 100 | ||
| CN218676.1 | - | 3 - 4 | 326 | 100 | ||
| CR406363.1 | - | 3 - 4 | 254 | 100 | ||
More Information
Set Membership: |
|

DNAJC5
Genomic Map