Assay Details
| Gene Symbol: DSCR3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015301454.2 | XP_015156940.1 | 6 - 7 | 786 | 98 | |
| XM_416728.5 | XP_416728.1 | 6 - 7 | 751 | 98 | ||
| GenBank mRNA | CR524457.1 | - | 6 - 7 | 737 | 98 | |
More Information
Set Membership: |
|

DSCR3
Genomic Map