Assay Details
| Gene Symbol: AQP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_424500.6 | XP_424500.3 | 4 - 5 | 563 | 83 | |
| GenBank mRNA | AB358970.1 | - | 3 - 4 | 342 | 83 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

AQP3| Gene Symbol: AQP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_424500.6 | XP_424500.3 | 4 - 5 | 563 | 83 | |
| GenBank mRNA | AB358970.1 | - | 3 - 4 | 342 | 83 | |
Set Membership: |
|