Assay Details
| Gene Symbol: AQP7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004937083.3 | XP_004937140.1 | 5 - 6 | 786 | 67 | |
| XM_004937085.3 | XP_004937142.1 | 5 - 6 | 732 | 67 | ||
| XM_015277432.2 | XP_015132918.1 | 4 - 5 | 727 | 67 | ||
| XM_015277433.2 | XP_015132919.1 | 4 - 5 | 728 | 67 | ||
| XM_015277435.2 | XP_015132921.1 | 4 - 5 | 411 | 67 | ||
| XM_025144776.1 | XP_025000544.1 | 4 - 5 | 730 | 67 | ||
| GenBank mRNA | AB359225.1 | - | 5 - 6 | 487 | 67 | |
More Information
Set Membership: |
|

AQP7
Genomic Map