Assay Details
| Gene Symbol: SHC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001293280.3 | NP_001280209.1 | 6 - 7 | 964 | 89 | |
| XM_004948266.3 | XP_004948323.1 | 6 - 7 | 2091 | 89 | ||
| XM_015298488.2 | XP_015153974.1 | 8 - 9 | 700 | 89 | ||
| XM_015298489.2 | XP_015153975.1 | 8 - 9 | 701 | 89 | ||
| XM_015298490.2 | XP_015153976.1 | 7 - 8 | 680 | 89 | ||
| XM_015298491.2 | XP_015153977.1 | 7 - 8 | 1924 | 89 | ||
| GenBank mRNA | AF035949.1 | - | 6 - 7 | 446 | 89 | |
| CR390625.1 | - | 6 - 7 | 456 | 89 | ||
More Information
Set Membership: |
|

SHC1
Genomic Map