Assay Details
| Gene Symbol: NME4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001282217.1 | NP_001269146.1 | 3 - 4 | 347 | 71 | |
| GenBank mRNA | AF043543.1 | - | 2 - 3 | 242 | 71 | |
| BG642102.1 | - | 3 - 4 | 339 | 71 | ||
| BG713316.1 | - | 3 - 4 | 347 | 71 | ||
| BG713347.1 | - | 3 - 4 | 331 | 71 | ||
| BX270330.4 | - | 1 - 2 | 104 | 71 | ||
More Information
Set Membership: |
|

NME4
Genomic Map