Assay Details
| Gene Symbol: CYB5R2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001271982.1 | NP_001258911.1 | 8 - 9 | 940 | 73 | |
| GenBank mRNA | AJ294706.1 | - | 4 - 5 | 366 | 73 | |
| AJ721007.1 | - | 8 - 9 | 935 | 73 | ||
| BU225498.1 | - | 4 - 5 | 354 | 73 | ||
More Information
Set Membership: |
|

CYB5R2
Genomic Map