Assay Details
| Gene Symbol: CREBRF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004944871.3 | XP_004944928.1 | 4 - 5 | 1489 | 66 | |
| GenBank mRNA | AJ719449.1 | - | 3 - 4 | 389 | 66 | |
More Information
Set Membership: |
|

CREBRF
Genomic Map