Assay Details
| Gene Symbol: TMEM208 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001256963.1 | NP_001243892.1 | 3 - 4 | 176 | 74 | |
| XM_015279105.2 | XP_015134591.1 | 2 - 3 | 78 | 74 | ||
| GenBank mRNA | AJ720252.1 | - | 3 - 4 | 191 | 74 | |
| BU296314.1 | - | 3 - 4 | 176 | 74 | ||
More Information
Set Membership: |
|

TMEM208
Genomic Map