Assay Details
| Gene Symbol: ADAMDEC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001233495.4 | XP_001233496.1 | 17 - 18 | 2007 | 60 | |
| XM_015297321.2 | XP_015152807.1 | 17 - 18 | 2007 | 60 | ||
| GenBank mRNA | AJ851685.1 | - | 16 - 17 | 1718 | 60 | |
More Information
Set Membership: |
|

ADAMDEC1
Genomic Map