Assay Details
| Gene Symbol: NDUFAB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004945263.3 | XP_004945320.1 | 3 - 4 | 517 | 142 | |
| GenBank mRNA | BX929522.1 | - | 3 - 4 | 472 | 142 | |
| BX934815.1 | - | 535 | 142 | |||
More Information
Set Membership: |
|

NDUFAB1
Genomic Map