Assay Details
| Gene Symbol: BORCS7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001277369.1 | NP_001264298.1 | 2 - 3 | 207 | 78 | |
| GenBank mRNA | AJ444511.1 | - | 2 - 3 | 326 | 78 | |
| BX929631.1 | - | 2 - 3 | 193 | 78 | ||
| BX932585.1 | - | 2 - 3 | 201 | 78 | ||
More Information
Set Membership: |
|

BORCS7
Genomic Map