Assay Details
| Gene Symbol: LNX1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004936026.3 | XP_004936083.1 | 3 - 4 | 914 | 88 | |
| XM_004936027.3 | XP_004936084.1 | 3 - 4 | 982 | 88 | ||
| XM_004936028.3 | XP_004936085.1 | 2 - 3 | 690 | 88 | ||
| XM_015285526.2 | XP_015141012.1 | 3 - 4 | 989 | 88 | ||
| XM_015285527.2 | XP_015141013.1 | 3 - 4 | 1001 | 88 | ||
| XM_015285528.2 | XP_015141014.1 | 3 - 4 | 813 | 88 | ||
| XM_015285529.2 | XP_015141015.1 | 3 - 4 | 874 | 88 | ||
| XM_015285530.2 | XP_015141016.1 | 3 - 4 | 981 | 88 | ||
| XM_015285531.2 | XP_015141017.1 | 3 - 4 | 986 | 88 | ||
| XM_420707.6 | XP_420707.2 | 2 - 3 | 754 | 88 | ||
| GenBank mRNA | BX931803.2 | - | 2 - 3 | 677 | 88 | |
More Information
Set Membership: |
|

LNX1
Genomic Map