Assay Details
| Gene Symbol: PTGES3L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001328354.1 | NP_001315283.1 | 2 - 3 | 207 | 108 | |
| XM_015299631.2 | XP_015155117.1 | 2 - 3 | 242 | 108 | ||
| XM_015299632.2 | XP_015155118.1 | 2 - 3 | 248 | 108 | ||
| XM_025143976.1 | XP_024999744.1 | 2 - 3 | 227 | 108 | ||
| GenBank mRNA | BX931852.2 | - | 2 - 3 | 180 | 108 | |
| BX933009.1 | - | 2 - 3 | 182 | 108 | ||
More Information
Set Membership: |
|

PTGES3L
Genomic Map