Assay Details
| Gene Symbol: MUM1L1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001282239.1 | NP_001269168.1 | 7 - 8 | 1761 | 102 | |
| GenBank mRNA | BU201616.1 | - | 4 - 5 | 336 | 102 | |
| BX930252.1 | - | 2 - 3 | 120 | 102 | ||
| BX933853.2 | - | 4 - 5 | 356 | 102 | ||
| BX936111.1 | - | 4 - 5 | 740 | 102 | ||
| CD727139.1 | - | 4 - 5 | 355 | 102 | ||
More Information
Set Membership: |
|

MUM1L1
Genomic Map