Assay Details
| Gene Symbol: MUM1L1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001282239.1 | NP_001269168.1 | 8 - 9 | 1821 | 80 | |
| GenBank mRNA | BU201616.1 | - | 5 - 6 | 396 | 80 | |
| BX930252.1 | - | 3 - 4 | 180 | 80 | ||
| BX933853.2 | - | 5 - 6 | 416 | 80 | ||
| BX936111.1 | - | 5 - 6 | 800 | 80 | ||
| CD727139.1 | - | 5 - 6 | 415 | 80 | ||
More Information
Set Membership: |
|

MUM1L1
Genomic Map