Assay Details
| Gene Symbol: MUM1L1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001282239.1 | NP_001269168.1 | 9 - 10 | 1894 | 118 | |
| GenBank mRNA | BU201616.1 | - | 6 - 7 | 469 | 118 | |
| BX930252.1 | - | 4 - 5 | 253 | 118 | ||
| BX933853.2 | - | 6 - 7 | 489 | 118 | ||
| CD727139.1 | - | 6 - 7 | 488 | 118 | ||
| CN218936.1 | - | 1 - 2 | 220 | 118 | ||
More Information
Set Membership: |
|

MUM1L1
Genomic Map