Assay Details
| Gene Symbol: MUM1L1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001282239.1 | NP_001269168.1 | 10 - 11 | 1952 | 81 | |
| GenBank mRNA | BU201616.1 | - | 7 - 8 | 527 | 81 | |
| BX930252.1 | - | 5 - 6 | 311 | 81 | ||
| BX933853.2 | - | 7 - 8 | 547 | 81 | ||
| CD727139.1 | - | 7 - 8 | 546 | 81 | ||
| CN218936.1 | - | 2 - 3 | 278 | 81 | ||
More Information
Set Membership: |
|

MUM1L1
Genomic Map