Assay Details
| Gene Symbol: MUM1L1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001282239.1 | NP_001269168.1 | 12 - 13 | 2473 | 122 | |
| GenBank mRNA | BU327777.1 | - | 2 - 3 | 376 | 122 | |
| BU435104.1 | - | 2 - 3 | 314 | 122 | ||
| BX930252.1 | - | 7 - 8 | 832 | 122 | ||
| BX933853.2 | - | 9 - 10 | 1068 | 122 | ||
| BX935774.2 | - | 1 - 2 | 466 | 122 | ||
| CR405834.1 | - | 1 - 2 | 565 | 122 | ||
| CR406265.1 | - | 1 - 2 | 565 | 122 | ||
More Information
Set Membership: |
|

MUM1L1
Genomic Map