Assay Details
| Gene Symbol: IFT27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001277380.1 | NP_001264309.1 | 2 - 3 | 228 | 103 | |
| NM_001277381.2 | NP_001264310.1 | 2 - 3 | 228 | 103 | ||
| XM_015285377.2 | XP_015140863.1 | 3 - 4 | 766 | 103 | ||
| XM_025148675.1 | XP_025004443.1 | 3 - 4 | 766 | 103 | ||
| GenBank mRNA | BU219304.1 | - | 2 - 3 | 139 | 103 | |
| BX934306.2 | - | 2 - 3 | 183 | 103 | ||
| BX935463.2 | - | 2 - 3 | 228 | 103 | ||
| CN226311.1 | - | 2 - 3 | 212 | 103 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015285377.2, XM_025148675.1 transcripts.
Set Membership: |
|

IFT27
Genomic Map