Assay Details
| Gene Symbol: MED31 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001321595.1 | NP_001308524.1 | 2 - 3 | 341 | 121 | |
| GenBank mRNA | BU240322.1 | - | 2 - 3 | 112 | 121 | |
| BX934309.2 | - | 2 - 3 | 115 | 121 | ||
| BX934600.2 | - | 2 - 3 | 361 | 121 | ||
More Information
Set Membership: |
|

MED31
Genomic Map