Assay Details
| Gene Symbol: PLPPR4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004936566.3 | XP_004936623.1 | 2 - 3 | 207 | 65 | |
| XM_015290740.2 | XP_015146226.1 | 3 - 4 | 1578 | 65 | ||
| XM_015290741.2 | XP_015146227.1 | 2 - 3 | 288 | 65 | ||
| XM_422319.5 | XP_422319.2 | 2 - 3 | 1343 | 65 | ||
| GenBank mRNA | BX950809.2 | - | 1 - 2 | 27 | 65 | |
More Information
Set Membership: |
|

PLPPR4
Genomic Map