Assay Details
| Gene Symbol: NUDT6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003641202.4 | XP_003641250.2 | 1 - 2 | 63 | 99 | |
| GenBank mRNA | CR388466.1 | - | 3 - 4 | 243 | 99 | |
More Information
Set Membership: |
|

NUDT6
Genomic Map