Assay Details
| Gene Symbol: OGFRL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015284795.2 | XP_015140281.1 | 6 - 7 | 824 | 87 | |
| XM_015284796.2 | XP_015140282.1 | 6 - 7 | 822 | 87 | ||
| GenBank mRNA | CR390319.1 | - | 4 - 5 | 368 | 87 | |
More Information
Set Membership: |
|

OGFRL1
Genomic Map